MilliporeSigma
  • Calmodulin interacts with ATP binding cassette transporter A1 to protect from calpain-mediated degradation and upregulates high-density lipoprotein generation.
Calmodulin interacts with ATP binding cassette transporter A1 to protect from calpain-mediated degradation and upregulates high-density lipoprotein generation.
Arteriosclerosis, thrombosis, and vascular biology (2010-04-17)
Noriyuki Iwamoto, Rui Lu, Nobukiyo Tanaka, Sumiko Abe-Dohmae, Shinji Yokoyama
ABSTRACT
To investigate the interaction of ATP-binding cassette transporter A1 (ABCA1) with calmodulin in relation to its calpain-mediated degradation because many calpain substrates bind calmodulin to regulate cellular functions. The activity of ABCA1 is regulated through proteolysis by calpain. An immunoprecipitation and glutathione S-transferase pull-down assay revealed that ABCA1 directly binds calmodulin in a Ca(2+)-dependent manner. The cytoplasmic loop of ABCA1 contains a typical calmodulin binding sequence of 1-5-8-14 motifs (1245 to 1257 amino acids). The peptide of this region showed binding to calmodulin, and deletion of the 1-5-8-14 motif abolished this interaction. This motif is located near the ABCA1 Pro-Glu-Ser-Thr sequence, and the presence of calmodulin/Ca(2+) protected the peptides from proteolysis by calpain. The knockdown of calmodulin by a specific small and interfering RNA increased the degradation of ABCA1 and decreased ABCA1 protein and apolipoprotein A-I-mediated lipid release. Surprisingly, calmodulin inhibitor W7 increased calmodulin binding to ABCA1 and protected it from calpain-mediated degradation, consistent with our previous finding that this compound increased apolipoprotein A-I-mediated cell cholesterol release. Calmodulin directly binds and stabilizes ABCA1 in the presence of Ca(2+) and increases the generation of high-density lipoprotein.