MilliporeSigma
Molecular cloning of murine FLT and FLT4.
Oncogene (1993-08-01)
H Finnerty, K Kelleher, G E Morris, K Bean, D M Merberg, R Kriz, J C Morris, H Sookdeo, K J Turner, C R Wood
ABSTRACT
A wide range of growth and differentiation processes are regulated by the signalling of receptor tyrosine kinases (RTKs). We have developed a nested polymerase chain reaction (PCR) procedure with degenerate primers, and used it to identify RTKs expressed in murine fetal thymus. A novel RTK, called FLT4, and the murine homologue of FLT were found, and their PCR fragment sequences were used to isolate larger cDNA clones spanning the complete coding regions of these receptors. FLT4 was found to contain an extracellular region similar to the corresponding sequences of FLT and Flk-1, containing seven immunoglobulin domains.