MilliporeSigma
  • The LIMD1 protein bridges an association between the prolyl hydroxylases and VHL to repress HIF-1 activity.
The LIMD1 protein bridges an association between the prolyl hydroxylases and VHL to repress HIF-1 activity.
Nature cell biology (2012-01-31)
Daniel E Foxler, Katherine S Bridge, Victoria James, Thomas M Webb, Maureen Mee, Sybil C K Wong, Yunfeng Feng, Dumitru Constantin-Teodosiu, Thorgunnur Eyfjord Petursdottir, Johannes Bjornsson, Sigurdur Ingvarsson, Peter J Ratcliffe, Gregory D Longmore, Tyson V Sharp
ABSTRACT
There are three prolyl hydroxylases (PHD1, 2 and 3) that regulate the hypoxia-inducible factors (HIFs), the master transcriptional regulators that respond to changes in intracellular O(2) tension. In high O(2) tension (normoxia) the PHDs hydroxylate two conserved proline residues on HIF-1α, which leads to binding of the von Hippel-Lindau (VHL) tumour suppressor, the recognition component of a ubiquitin-ligase complex, initiating HIF-1α ubiquitylation and degradation. However, it is not known whether PHDs and VHL act separately to exert their enzymatic activities on HIF-1α or as a multiprotein complex. Here we show that the tumour suppressor protein LIMD1 (LIM domain-containing protein) acts as a molecular scaffold, simultaneously binding the PHDs and VHL, thereby assembling a PHD-LIMD1-VHL protein complex and creating an enzymatic niche that enables efficient degradation of HIF-1α. Depletion of endogenous LIMD1 increases HIF-1α levels and transcriptional activity in both normoxia and hypoxia. Conversely, LIMD1 expression downregulates HIF-1 transcriptional activity in a manner depending on PHD and 26S proteasome activities. LIMD1 family member proteins Ajuba and WTIP also bind to VHL and PHDs 1 and 3, indicating that these LIM domain-containing proteins represent a previously unrecognized group of hypoxic regulators.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.