MilliporeSigma
  • A Lin-9 complex is recruited by B-Myb to activate transcription of G2/M genes in undifferentiated embryonal carcinoma cells.
A Lin-9 complex is recruited by B-Myb to activate transcription of G2/M genes in undifferentiated embryonal carcinoma cells.
Oncogene (2009-03-03)
A S Knight, M Notaridou, R J Watson
ABSTRACT
It has recently been discovered that cell-cycle gene transcription is regulated by a core complex named LINC that switches from a transcriptionally repressive complex in G(0)-G(1) with the p130 or p107 pocket proteins and E2F4 to a transcriptionally active complex in S-G(2) containing B-Myb. We have studied the function of LINC in F9 embryonal carcinoma cells, which are distinguished by a rapid cell cycle resulting from an extremely short G(1) phase. We show that suppressing expression of the LINC component, Lin-9, in F9 cells causes arrest in mitosis, and we have used this system to screen for transcriptional targets. In these cells, B-Myb was found in complexes with Lin-9 and several other LINC constituents, however, the pocket proteins did not associate with LINC unless F9 cells were differentiated. Lin-9 and B-Myb were both required for transcription of G(2)/M genes such as Cyclin B1 and Survivin. Moreover, B-Myb was demonstrated to recruit Lin-9 to the Survivin promoter through multiple Myb-binding sites. The demonstration that a B-Myb/LINC complex is vital for progression through mitosis in cells lacking a G(1)/S checkpoint has implications for both undifferentiated embryonal cells and for cancers in which pocket protein function is compromised.