MilliporeSigma
  • Zinc finger domain of Snail functions as a nuclear localization signal for importin beta-mediated nuclear import pathway.
Zinc finger domain of Snail functions as a nuclear localization signal for importin beta-mediated nuclear import pathway.
Genes to cells : devoted to molecular & cellular mechanisms (2005-04-20)
Hideki Yamasaki, Toshihiro Sekimoto, Tadashi Ohkubo, Tsutomu Douchi, Yukihiro Nagata, Masayuki Ozawa, Yoshihiro Yoneda
ABSTRACT
Snail, a DNA-binding zinc finger protein, functions as a transcriptional repressor for genes including E-cadherin during development and the acquisition of tumor cell invasiveness. Human Snail is a 264-amino acid nuclear protein with an amino-terminal basic amino acid-rich domain (SNAG domain) and a carboxyl-terminal DNA-binding domain (zinc finger domain). A series of fusion proteins composed of green fluorescent protein (GFP) and portions of the Snail protein were generated, and their subcellular localization was examined. Fusion of the four zinc fingers to GFP led to the targeting of GFP to the nucleus, demonstrating that the zinc finger domain is sufficient for nuclear localization. Using an in vitro transport system, the nuclear import of Snail was reconstituted by importin (karyopherin) beta in the presence of Ran and NTF2. We further demonstrated that Snail binds directly to importin beta in a zinc finger domain-dependent manner. These results indicate that zinc finger domain of Snail functions as a nuclear localization signal and Snail can be transported into the nucleus in an importin beta-mediated manner.