MilliporeSigma
  • Focal cerebral ischemia in rats alters APP processing and expression of Abeta peptide degrading enzymes in the thalamus.
Focal cerebral ischemia in rats alters APP processing and expression of Abeta peptide degrading enzymes in the thalamus.
Neurobiology of disease (2009-05-12)
Mikko Hiltunen, Petra Mäkinen, Sirpa Peräniemi, Juhani Sivenius, Thomas van Groen, Hilkka Soininen, Jukka Jolkkonen
ABSTRACT
We have previously demonstrated aggregation of amyloid precursor protein (APP) and beta-amyloid (Abeta) to dense plaque-like deposits in the thalamus of rats subjected to transient middle cerebral artery occlusion (MCAO). Here, we investigated the underlying molecular effects of MCAO on APP processing and expression profiles of Abeta degrading enzymes in the cortex adjacent to the infarct (penumbra) and ipsilateral thalamus 2, 7 and 30 days after ischemic insult. Enhanced beta-amyloidogenic processing of APP and altered insulin degrading enzyme and neprilysin expression were observed in the thalamus, but not the penumbral cortex, 7 and 30 days after MCAO coinciding with increased calcium levels and beta-secretase (BACE) activity. Consecutively, increased BACE activity associated with depletion of BACE trafficking protein GGA3, suggesting a post-translational stabilization of BACE. These results demonstrate that focal cerebral ischemia leads to complex pathogenic events in the thalamus long after the initial insult.