MilliporeSigma
  • Agrin is involved in lymphocytes activation that is mediated by alpha-dystroglycan.
Agrin is involved in lymphocytes activation that is mediated by alpha-dystroglycan.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2006-01-06)
Jinping Zhang, Ying Wang, Yiwei Chu, Liping Su, Yanping Gong, Ruihua Zhang, Sidong Xiong
ABSTRACT
It is well established that agrin, an extracellular matrix protein, plays a crucial role in the formation of neuromuscular junctions. Recent evidence indicates that agrin also contributes to immunological synapse formation. However, little is known about how agrin induces the activation of lymphocytes and whose receptors mediate its regulatory effects on these cells. In the present study, agrin was detected in lymphocytes. Up-regulation of agrin expression was involved in lymphocyte activation whereas down-regulation of its expression led to inhibition of both antigen-specific and nonspecific lymphocyte activation, indicating an intrinsic role for agrin in the activation of lymphocytes. Unexpectedly, unlike that found in muscle cells where there is coexpression of muscle-specific kinase (MuSK) and alpha-dystroglycan receptors for agrin, only alpha-dystroglycan could be detected in lymphocytes. Confocal examination showed that alpha-dystroglycan colocalized with agrin in forming the immunological synapse. Down-regulation of alpha-dystroglycan expression inhibited lymphocyte activation even in the presence of agrin. However, agrin involved in down-regulation of alpha-dystroglycan receptors did not increase the inhibitory effect on lymphocytes activation. The anti-alpha-dystroglycan antibody also induced lymphocytes activation. Taken together, these findings strongly indicate that agrin and alpha-dystroglycan mediate lymphocyte activation. Furthermore, agrin-involved lymphocyte activation is mediated by alpha-dystroglycan.