MilliporeSigma
  • Immunolabeling reveals cellular localization of the N-methyl-D-aspartate receptor subunit NR2B in neurosecretory cells but not astrocytes of the rat magnocellular nuclei.
Immunolabeling reveals cellular localization of the N-methyl-D-aspartate receptor subunit NR2B in neurosecretory cells but not astrocytes of the rat magnocellular nuclei.
The Journal of comparative neurology (2000-10-24)
M C Currás-Collazo, C Chin, G Díaz, C Stivers, L Bozzetti, L Y Tran, G Kíaz
ABSTRACT
Previous studies suggest that activation of N-methyl-D-aspartate (NMDA) receptors facilitates phasic firing and spike clustering displayed by magnocellular neuroendocrine cells (MNCs) of the supraoptic (SON) and paraventricular nucleus of the hypothalamus (PVN). Osmotic stimulation produces similar activity patterns which, in turn, can lead to enhanced release of vasopressin and oxytocin from MNCs. Our laboratory has shown that dehydration regulates the expression of the NMDA receptor subunits, NR1 and NR2B, in the SON and PVN, suggesting their involvement in osmoregulation. In the present study, we examined the cellular localization of NR2B, one of the glutamate-binding subunits of the NMDA receptor, with an NR2B-specific antibody. Using double-label immunohistochemistry and three different detection methods with metallic, peroxidase, and fluorescence markers, it was found that both vasopressin and oxytocin-producing MNC populations synthesize NR2B. The incidence of NR2B colocalization with vasopressin-neurophysin in the SON and lateral magnocellular PVN (PVL) was 0.95 and 0.91, respectively. For oxytocin-neurophysin, the corresponding values were 0.97 and 0.95, respectively. Furthermore, the extent of colocalization in MNCs of the SON, PVL, retrochiasmatic SON, and accessory neurosecretory nuclei was similar. Astrocytes associated with the SON, and identified with antibodies targeting glial fibrillary acidic protein (GFAP) or vimentin, were not colabeled with NR2B. Our results demonstrate that NR2B protein is expressed by almost all MNCs and that it is equally prevalent in vasopressinergic and oxytocinergic populations of various magnocellular neuroendocrine nuclei supporting a role of NMDA receptors in MNC-mediated neurosecretory processes. Although NR2B may form part of functional NMDA receptors on MNCs, it is probably not present on astrocytes associated with nearby MNCs.