MilliporeSigma
  • N-methyl-D-aspartate receptor subunit NR2B is widely expressed throughout the rat diencephalon: an immunohistochemical study.
N-methyl-D-aspartate receptor subunit NR2B is widely expressed throughout the rat diencephalon: an immunohistochemical study.
The Journal of comparative neurology (2000-11-14)
A M Khan, B G Stanley, L Bozzetti, C Chin, C Stivers, M C Currás-Collazo
ABSTRACT
Glutamate (Glu), a major excitatory neurotransmitter within the hypothalamus and thalamus, acts upon many receptors, including the N-methyl-D-aspartate (NMDA) subtype. Abundant evidence suggests that variations in the subunit composition of NMDA receptors (NMDA-Rs) contribute to differences in Glu's immediate electrophysiological effects as well as to the patterns of signal transduction cascades it triggers to mediate long-term changes in neuronal function. We have previously shown that hypothalamic NMDA-Rs containing the NR2B subunit may be involved in the control of eating as well as in the mediation of physiological responses to osmotic stimuli. To broaden our understanding of diencephalic NMDA-R participation in other functions, we localized the NR2B subunit in the diencephalon of the adult male rat using immunoperoxidase, immunogold, and immunofluorescence techniques and an affinity-purified polyclonal antibody specific for the NR2B subunit of the NMDA-R. In addition, we used a monoclonal NR2B antibody with immunoperoxidase detection to confirm the NR2B distribution seen with the polyclonal antibody. In the hypothalamus, the highest levels of NR2B immunoreactivity (-ir) were found in the magnocellular neurosecretory system, including the paraventricular and supraoptic nuclei. A new finding was that intense NR2B-ir was present within perivascular "accessory" magnocellular groups of this system, including the nucleus circularis, anterior fornical nucleus, and scattered clusters of lateral hypothalamic cells apposed to blood vessels. Robust NR2B-ir was also present within the arcuate nucleus, the median eminence, and the tuberal nucleus, and light immunostaining was found in all other hypothalamic nuclei examined. In the thalamus, the highest NR2B-ir was observed in the medial habenula and the anterodorsal, paraventricular, rhomboid, reticular, and dorsal lateral geniculate nuclei. As in the hypothalamus, all thalamic nuclei examined displayed at least light immunostaining for this subunit. Control sections, including those incubated with the polyclonal NR2B antibody preadsorbed with its fusion protein, were virtually devoid of immunostaining. This demonstration that the NR2B subunit of the NMDA-R is widely distributed in the diencephalon, implicates it in a wide variety of functions, and provides a useful anatomical framework for establishing a comprehensive map of Glu receptor populations within this major subdivision of the brain.