MilliporeSigma
Formation of Phenolic Compounds from d-Galacturonic Acid.
Journal of agricultural and food chemistry (2018-10-09)
Alexandra Urbisch, Ulrike Einhorn-Stoll, Hanna Kastner, Stephan Drusch, Lothar W Kroh
ABSTRACT
Aqueous d-galacturonic acid (d-GalA) model systems treated at 130 °C at different pH values show an intense color formation, whereas other reducing sugars, such as d-galactose (d-Gal), scarcely react. GC-MS measurements revealed the presence of several phenolic compounds: e.g., 3,8-dihydroxy-2-methyl-4 H-chromen-4-one (chromone) and 2,3-dihydroxybenzaldehyde (2,3-DHBA). These phenolic compounds, especially 2,3-DHBA, possess an intense browning potential and cannot be found within heated model solutions of reducing sugars. Investigations regarding the formation of these substances show that α-ketoglutaraldehyde plays an important role as an intermediate product. In addition, MS analysis of model systems of norfuraneol in combination with 2,3-DHBA showed the formation of oligomers that could also be detected in d-GalA model systems, leading to the assumption that, in addition to reductic acid, these compounds are jointly responsible for the strong color formation during the heat treatment of d-GalA.
Sigma-Aldrich
D-(+)-Galacturonic acid monohydrate, ≥97% (titration)