MilliporeSigma
  • Idarubicin-145-2C11-F(ab')2 promotes peripheral tolerance and reduces chronic vascular disease in mouse cardiac allografts.
Idarubicin-145-2C11-F(ab')2 promotes peripheral tolerance and reduces chronic vascular disease in mouse cardiac allografts.
Transplant immunology (2000-01-19)
W R Han, L J Murray-Segal, A Gershenzon, J G Zhang, A N Hodder, G A Pietersz, P L Mottram
ABSTRACT
In order to reduce the toxic effects of the T cell activating anti-CD3 monoclonal antibody, 145-2C11, F(ab')2 fragments were prepared by pepsin digestion. These fragments were then used as non-immunosuppressive carriers for the cytotoxic drug idarubicin (IDA), to reduce toxicity of both the monodonal antibodies (mAb) and the drug and to increase the specificity of drug delivery. The IDA-145-2C11 F(ab')2 immunoconjugate was tested for specificity by fluorometry. 145-2C11 intact antibody, 145-2C11 F(ab')2 and IDA conjugates of the antibody and F(ab')2 were used to treat CBA recipients of BALB/c vascularized cardiac allografts. Mice with hearts surviving >100 days were challenged with donor and third party (C57BL/6) skin grafts. Although both antibody and F(ab')2 blocked the binding of 145-2C11-FITC to CBA spleen cells, only the intact antibody caused sustained depletion of CD3 cells in vivo. 145-2C11 F(ab')2 blocked cell surface CD3 within 30 min, but was cleared in 24 h without depletion of CD3 cells from the spleen. In BALB/c to CBA cardiac allografts (rejected in 12-17 days), IDA-145-2C11 F(ab')2 (0.2 mg/20 g mouse i.p. at the time of transplantation) induced >100 days' allograft survival and specific tolerance, in contrast to the equivalent dose of 145-2C11 F(ab')2 (mean survival 25 days). Hearts from IDA-145-2C11 F(ab')2-treated mice at >100 days showed decreased cellular infiltration and less chronic vascular disease than long-surviving hearts from mice treated with an alternative antibody, KT3. Thus, F(ab')2 prepared from 145-2C11 provided a suitable CD3-specific, nonimmunosuppressive carrier for IDA. This immunoconjugate was more effective against both acute and chronic rejection than other conjugates or whole antibody. IDA-145-2C11 F(ab')2 is an effective, nontoxic tolerogen in the mouse cardiac allograft model.