Sand
Synonym(s):
Sand, Sand, white quartz, Quartz, Silicon dioxide
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing