Skip to Content
MilliporeSigma
Pyoktanin Blue SAJ special grade

Pyoktanin Blue

Pyoktanin Blue SAJ special grade
Synonym(s):
Methyl violet 2B, Basic Violet 1
CAS Number:

Products

Brand
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing