Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8278

Supelco

Carbinoxamine maleate salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2O · C4H4O4
CAS Number:
Molecular Weight:
406.86
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCOC(c1ccc(Cl)cc1)c2ccccn2

InChI

1S/C16H19ClN2O.C4H4O4/c1-19(2)11-12-20-16(15-5-3-4-10-18-15)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-10,16H,11-12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

GVNWHCVWDRNXAZ-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Rachelefsky et al.
The Journal of allergy and clinical immunology, 69(4), 382-387 (1982-04-01)
We evaluated the role of antimicrobials in the treatment of chronic maxillary sinusitis in children with respiratory allergy. Night and day cough, nasal obstruction, rhinorrhea, postnasal seen. Eighty-four children were treated in a double-blind manner with either amoxicillin, erythromycin, trimethoprim-sulfamethoxazole
J T Connell et al.
Drug intelligence & clinical pharmacy, 18(3), 244-247 (1984-03-01)
In a double-blind study, a timed-release tablet containing carbinoxamine maleate 8 mg and pseudoephedrine hydrochloride 120 mg was compared with placebo for the treatment of signs and symptoms of nasal allergy. Ninety-four adults with rhinitis caused by grass or ragweed
A Stockis et al.
Arzneimittel-Forschung, 45(9), 1009-1012 (1995-09-01)
The plasma pharmacokinetics of carbinoxamine (CA, CAS 486-16-8) and phenylephrine (PE, CAS 59-42-7) after single dose administration of a retard capsule (Rhinopront) containing 20 mg PE hydrochloride and 4 mg CA maleate, were compared to those of the same active
A Stockis et al.
Arzneimittel-Forschung, 42(12), 1478-1481 (1992-12-01)
The plasma pharmacokinetics of carbinoxamine (CA, CAS 486-16-8) and phenylpropanolamine (PP, CAS 14838-15-4) after single dose administration of a retard suspension (Rhinopront), containing a resinate as sustained-release agent, were compared to those of the same active principles given as an
N J Pérez Carmona et al.
Revista alergia Mexico (Tecamachalco, Puebla, Mexico : 1993), 44(3), 70-73 (1997-05-01)
They were studied 48 children of 3 to 8 years old, of two sex (30 male and 18 female), that attended the external service of otolaryngology of the Hospital of the ISSSTE of Nuevo Laredo, Tamaulipas (Mexico) and the private

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service