70382
Sodium 1,2-naphthoquinone-4-sulfonate
for spectrophotometric det. of isonicotic hydrazide, amines and amino acids, ≥97.0% (T)
Synonym(s):
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 3,4-Dihydro-3,4-dioxo-1-naphthalenesulfonic acid sodium salt, Folin’s reagent
About This Item
Recommended Products
assay
≥97.0% (HPLC)
≥97.0% (T)
form
powder
quality
for spectrophotometric det. of isonicotic hydrazide, amines and amino acids
specific analyte(s)
sodium
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
solubility
H2O: 50 mg/mL, clear
SMILES string
[Na+].[O-]S(=O)(=O)C1=CC(=O)C(=O)c2ccccc12
InChI
1S/C10H6O5S.Na/c11-8-5-9(16(13,14)15)6-3-1-2-4-7(6)10(8)12;/h1-5H,(H,13,14,15);/q;+1/p-1
InChI key
UBLXEEBHYISRFM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service