EZFITH0LD3
EZ-Fit® Manifold
3-place, for rubber stopper tulip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel
sterilization compatibility
autoclavable compatible
manufacturer/tradename
EZ-Fit®
input
sample type bottled water
sample type pharmaceutical(s)
sample type cosmetics
sample type beer
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
Related Categories
General description
EZ-Fit® Manifold′s unique design optimizes workflow and maintenance and reduces the risk of contamination. Different filtration heads fit both reusable and disposable filtration devices. Low height design increases comfort when used in laminar flow hoods. EZ-Fit® Manifold prevents biofilm formation because every single component can be removed for easy cleaning and autoclaving.
Application
The EZ-Fit® Manifold is a filtration manifold used for universal laboratory filtrations. It supports the simultaneous filtration of three test samples. The manifold can be used with EZ-Fit® filtration units, Microfil® filtration system, 55-Plus Monitor, and standard glass or stainless-steel funnels.
Legal Information
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
Configured for
Product No.
Description
Pricing
Related product
Product No.
Description
Pricing
accessory
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service