Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09769

Supelco

Case

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311
NACRES:
SE.11

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 30
for use with Spectroquant® Nova 60

storage temp.

2-30°C

General description

NOVA 30, NOVA 60 Spectroquant®
This rugged case is ideal for transporting a Spectroquant NOVA 30, NOVA 60 or NOVA 60A (not included). It features a foam inlay that idealy protects the instrument. Therefore it can be used to transport the device in the field to perform e.g. water analysis directly on site.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service