Key Documents
Z171387
Supelco
Series A-2 gas syringe
capacity 25 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
2 in.
feature
needle bevel tip
manufacturer/tradename
VICI 050023
capacity
25 μL
needle L
2 in.
General description
Unique Sample-Lok push-button valve allows sample storage in syringes as small as 25μL and permits pressurization of samples prior to injection, for better resolution. Plunger, when fully depressed, makes flush contact with bottom of syringe barrel, virtually eliminating dead volume. Parts are replaceable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)