Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS009189

Sigma-Aldrich

4-Methyl-4-mercaptopentan-2-one

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12OS
CAS Number:
Molecular Weight:
132.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CC(=O)CC(C)(C)S

InChI

1S/C6H12OS/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3

InChI key

QRNZMFDCKKEPSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.2 °F

flash_point_c

54 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alvaro Lezaeta et al.
Food research international (Ottawa, Ont.), 108, 595-603 (2018-05-08)
We developed a sensory-based methodology to aromatically enrich wines using different aromatic fractions recovered during fermentations of Sauvignon Blanc must. By means of threshold determination and generic descriptive analysis using a trained sensory panel, the aromatic fractions were characterized, selected
Nikolaos Kontoudakis et al.
Food research international (Ottawa, Ont.), 98, 95-102 (2017-06-15)
A variety of techniques have been developed with the ability to measure different forms of metals in wine with the ultimate aim of providing a more accurate indicator of metal induced spoilage of wine. This study was conducted in order
Sílvia Petronilho et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
Wine aroma is the result of complex interactions between volatile compounds and non-volatile ones and individual perception phenomenon. In this work, an aroma network approach, that links volatile composition (chromatographic data) with its corresponding aroma descriptors was used to explain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service