Kluczowe dokumenty
Z740017
BRAND® filter tips, racked, TipRack
volume 1-20 μL, BIO-CERT®
About This Item
Kod UNSPSC:
41121601
NACRES:
NB.01
maksymalna pojemność:
20 μL
pojemność:
1-20 μL
sterylność:
sterile; autoclaved
Właściwości:
RNase and DNase free
endotoxin free
filter
BIO-CERT®
opakowanie:
pack of 960 ea (10 racks of 96)
producent / nazwa handlowa:
BRAND 732626
Materiały
colorless
colorless polypropylene tip and filter element (polyethylene)
sterylność
sterile; autoclaved
Właściwości
RNase and DNase free
endotoxin free
filter
BIO-CERT®
opakowanie
pack of 960 ea (10 racks of 96)
producent / nazwa handlowa
BRAND 732626
maksymalna pojemność
20 μL
pojemność
1-20 μL
Opis ogólny
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. These filters function with consistent reliability. On the other hand, liquids can pass very slowly should they accidentally contact the filter.
Since the filter does not swell as in the case of self-sealing filter tips, the sample can be recovered from the filter by simply actuating the pipette′s blow-out function, or by centrifugation if necessary. This is clearly an important advantage of non-self-sealing filters, especially when working with valuable samples. As an added advantage, the absence of filter additives protects samples from this source of contamination.
NEW! TipRack, non-sterile and sterile, BIO-CERT®
Space-saving refill unit for TipBox. Protected in an environmentally compatible packaging of recyclable PET. Compared to the filled TipBoxes, the amount of waste with the new refill units is reduced by 20%. All pipette tips and filter tips up to 1000 μL are free of DNA, RNases, endotoxins and ATP. TipRacks in BIO-CERT® quality are sterile according to ISO 11 137 and the AAMI guidelines, a SAL of 10-6 is obtained.
These racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilized TipBox. All tip-trays are printed on one side with information on the contents.
Cechy i korzyści
• Cleanroom quality for contamination-free analyses.• Perfect fit for the Transferpette® micropipette and many other pipettes.• The tip system for all requirements.
Inne uwagi
BR732626 (India)
Informacje prawne
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)