About This Item
Kod UNSPSC:
23151806
eCl@ss:
32031690
Polecane produkty
Poziom jakości
Powiązane kategorie
Opis ogólny
Fittings Kit for the Viresolve® Pro and Pro+ Magnus Holder.
3 x 3.81 cm (1.5 in.) sanitary fittings
2 x 3.81 cm (1.5 in.) blanks
1 x 1.27 cm (.5 in.) vent fitting
1 x 1.27 cm (.5 in.) blank
3 x 3.81 cm (1.5 in.) sanitary fittings
2 x 3.81 cm (1.5 in.) blanks
1 x 1.27 cm (.5 in.) vent fitting
1 x 1.27 cm (.5 in.) blank
Komponenty
Kit includes: three 1.5 in. sanitary fittings, two 1.5 in. blanks, one vent fitting and one vent blank
Informacje prawne
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany