Kluczowe dokumenty
TQNXA4RP1
Millipore
Steritest® Equinox Essential Requalification Protocol
A4 format
About This Item
Kod UNSPSC:
41116500
NACRES:
NB.79
kompatybilność
for use with Steritest® Equinox pump.
Opis ogólny
Steritest® Equinox Essential Requalification protocol is designed as a ready to use guide to perform the requalification of the Steritest® Equinox pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals.
Zastosowanie
Steritest® Equinox Essential Requalification protocol is used for the re-qualification of Steritest® Equinox pump which is used to perform sterility testing for pharmaceutical products.
Informacje prawne
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany