Kluczowe dokumenty
Z566136
Pigment-free, filler-free sleeve stopper septa
bottom I.D. × O.D. 4.2 mm × 10.7 mm
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.35
Polecane produkty
Materiały
amber septa
natural rubber septa
Połączenie
joint ST/NS10/30
Właściwości
Use with small tubes
dno śr. wewn. × śr. zewn.
4.2 mm × 10.7 mm
Opis ogólny
These sleeve stopper septa are manufactured from pure natural rubber without fillers or pigments for super tactility and resealability. Less additives in the rubber lowers the potential for contamination during use.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z566136-100EA | 4061832795591 |