Z217514
Flexible stirrer shaft coupling
for glass shaft O.D. 10 mm, for motor shaft O.D. 1/4 in.
About This Item
Kod UNSPSC:
41100000
NACRES:
NB.72
Polecane produkty
Powiązane kategorie
Opis ogólny
Flexible coupling allows angular and parallel misalignment of glass stirring shaft without breakage. Neoprene rubber sleeve body absorbs shock and vibration and has great torsional stiffness for quiet, smooth running. PTFE sleeve slides over end of glass shaft. SS set screw secures other end of coupling to motor shaft.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z217514-1EA | 4061833308011 |