Z183954
Aldrich® large-capacity cold trap
Inside finger for traps with 50mm O-ring
About This Item
Kod UNSPSC:
41104812
NACRES:
NB.43
Polecane produkty
Powiązane kategorie
Opis ogólny
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z183954-1EA | 4061826227848 |