Kluczowe dokumenty
Y907200
Sigma-Aldrich
Valve
brass diaphragm, for CGA 540 right angle path, internal, right hand thread
About This Item
Kod UNSPSC:
12142200
NACRES:
NA.12
Materiały
brass diaphragm
Parametry
2640 psi max. pressure
Długość
4.62 in.
masa
628 g
dopasowywanie
for CGA 540 right angle path, internal, right hand thread
Inne uwagi
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)