Kluczowe dokumenty
CLS615969L
PYREXPLUS® coated solution bottles
capacity 9.5 L
Synonim(y):
Corning® 61596
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.43
pojemność:
9.5 L
producent / nazwa handlowa
Corning 61596-9L
pojemność
9.5 L
Opis ogólny
Features a protective plastic coating that helps prevent glass from shattering and reduces any spills. Fully autoclavable (121°C) and resistant to thermal shock. Graduated in 500mL increments with black enamel. Uses rubber stopper no. 12.
Informacje prawne
Corning is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)