GF50091320
Nickel
tube, 1000mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, hard, 99.5%
Synonym(s):
Nickel, NI007300
About This Item
Assay
99.5%
form
tubes
manufacturer/tradename
Goodfellow 500-913-20
resistivity
6.97 μΩ-cm, 20°C
L × wall thickness
1000 mm × 0.2 mm
O.D. × I.D.
2.0 mm × 1.6 mm
bp
2732 °C (lit.)
mp
1453 °C (lit.)
density
8.9 g/mL at 25 °C (lit.)
SMILES string
[Ni]
InChI
1S/Ni
InChI key
PXHVJJICTQNCMI-UHFFFAOYSA-N
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Skin Sens. 1 - STOT RE 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
SKU | GTIN |
---|---|
GF50091320-1EA | 4061825401539 |
GF50091320-2EA | 4061829500450 |
GF50091320-5EA | 4061829569341 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service