Key Documents
AXYAM384PCRRD
Sealing mat
for PCR microplates, size 384 wells, pkg of 5x10mats/cs
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122410
NACRES:
NB.22
material
polypropylene
무균
non-sterile
제조업체/상표
Axygen Inc AM-384-PCR-RD
Corning
포장
pkg of 5x10mats/cs
크기
384 wells
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
  • Axygen® sealing films are used in PCR storage applications.
  • Sealing films offer high-temperature resistance.
  • Made from autoclavable research-grade silicone
  • Reusable - following a bleach wash and ethanol rinse protocol.
법적 정보
Axygen is a registered trademark of Corning, Inc.