주요 문서
Z742110
Sigma-Aldrich
KitAlysis 24-well reaction block replacement film
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.51
Quality Level
일반 설명
KitAlysis 24-well Reaction Block replacement films are for use with the KitAlysis 24-well Reaction Block (Z742107) as a part of the KitAlysis High-Throughput Screening Platform. Replacement films are designed for single use and should be replaced between runs for optimal results.
KitAlysis 24-well Reaction Block required for operation (sold separately). Best when used with KitAlysis Benchtop Inertion Box (sold separately).
법적 정보
6538 is a trademark of American Type Culture Collection
KitAlysis is a trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)