일반 설명
For use with Aldrich products packed in Sure/Seal bottles. Screws over Sure/Seal crown cap to permit repeated dispensing of product via syringe while ensuring positive valved closure. Technical Information Bulletin AL-195, includes instructions for use of the valve.
법적 정보
Oxford is a registered trademark of Oxford Worldwide LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
이미 열람한 고객
Global Trade Item Number
SKU | GTIN |
---|---|
Z406260-1EA | 4061826667712 |