Merck
Z741961
Cameo Syringe Filter
PVDF membrane, pore size 0.22 μm, diam. 30 mm
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41104900
NACRES:
NB.24
ポアサイズ:
0.22 μm
直径:
30 mm
無菌性:
non-sterile
物質:
PVDF membrane
polypropylene housing
包装:
pkg of 50 ea
物質
PVDF membrane
polypropylene housing
無菌性
non-sterile
包装
pkg of 50 ea
メーカー/製品名
GVS Life Sciences 3038551
直径
30 mm
ポアサイズ
0.22 μm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
法的情報
Cameo is a trademark of GVS S.p.A.
最新バージョンのいずれかを選択してください:
試験成績書(COA)