Merck
Z672947
BRAND® accu-jet®プロ シリコーン製交換用アダプター
valve L × W × H 2 in. × 0.5 in. × 0.5 in., PTFE valve (non-return)
別名:
Tubing Connectors
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
物質
PTFE valve (non-return)
メーカー/製品名
BRAND 26508
包装
1 ea of
バルブL × W × H
2 in. × 0.5 in. × 0.5 in.
詳細
The BRAND® accu-jet® pro silicone pipette adapter features a reliable PTFE check valve, ensuring efficient liquid transfer without backflow. Compact in size (L 2" x W 0.5" x H 0.5"), this non-return valve replacement adapter enhances the functionality of your pipette while providing a secure fit.
特徴および利点
  • Silicone pipette adapter with PTFE check valve
  • Dimensions: L 2" x W 0.5" x H 0.5"
  • Non-return valve
その他情報
BR26508 (India)
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)