Merck
HS120254
Wide mouth safety labelled wash bottle
capacity 500 mL, label, Methanol, PE-LD
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41103209
NACRES:
NB.43
物質
PE-LD
green bottle
low-density polyethylene bottle
polypropylene cap
wide-mouth bottle
特徴
label Methanol
vented
with cap
包装
pack of 6 ea
メーカー/製品名
Heathrow Scientific HS120254
キャパシティ
500 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Clear and informative product safety information.
The information provided on the front of each bottle is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling′ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • With the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection and has a HDPE closure, the bottles are made from LDPE, with a PP closure
  • Six named chemicals, a self labeling and assorted pack of bottles to choose from
  • 500 mL bottle size
  • Supplied in packs of 6
Self Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
最新バージョンのいずれかを選択してください:
試験成績書(COA)