Merck
HS120099
Pearl Pipette
adjustable, volume 2-20 μL, light purple, single-channel
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
物質
light purple
white
特徴
adjustable
single-channel
包装
pack of 1 ea
メーカー/製品名
Heathrow Scientific HS120099
容積測定範囲
2-20 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
特徴および利点
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
法的情報
Pearl is a trademark of Specac, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)