Merck
GE18-1442-01
Sigma-Aldrich
HR 16 Packing Equipment
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41115700
NACRES:
SB.52
詳細
Packing equipment HR 16 is intended for the efficient packing of Cytiva High Resolution (HR) columns, with a diameter of 16 mm. The connector 16/16 connects your HR 16 column to the packing reservoir, the chromatografic tube HR 16/30. The preflanged tubing is connected to the inner end piece. The chromatographic tube seals against the inner end piece, and the protective jacket is held in position by the connector 16/16 and the outer end piece. The materials of the packing consist of fluoroplastics, fluororubber, and glass.
最新バージョンのいずれかを選択してください:
試験成績書(COA)