Merck
EP951020573
Eppendorf® twin.tec PCR Plates 384
Optimized rim for barcodes, PCR clean, colorless, pack of 300 ea
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41106300
物質
colorless
conical bottom
polypropylene wells
無菌性
non-sterile
特徴
PCR clean
lid: no
skirt
包装
pack of 300 ea
メーカー/製品名
Eppendorf® 951020573
サイズ
384 well
ウェル容積
45 μL
適合性
suitable for PCR
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
  • Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, blue, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
特徴および利点
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
法的情報
Eppendorf is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)