Merck
EP4926000042
Eppendorf® Reference® 2 Variable Volume Pipettor
12-channel, incl. epT.I.P.S.® Box, volume 10-100 μL, yellow
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
物質
yellow
特徴
12-channel
mechanical
包装
pack of 1 ea
メーカー/製品名
Eppendorf® 4922000048 (Global)
容量
10-100 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
法的情報
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)