特徴
for 45L CO2 Incubators (H3565-45 only)
包装
pack of 1 ea
メーカー/製品名
Benchmark Scientific H3565-45-LCI
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The (patent pending) IncuView LCI (Live Cell Imaging) technology for viewing cells in “real time” is an internal microscope and large, external display that allows cells to be viewed without removal from the incubator, eliminating the inherent temperature fluctuations and risk of contamination. Cells are easily viewed on the large, integrated display or viewed locally or remotely with a wifi enabled phone, tablet or computer. Cell viewing requires the IncuView Integrated Microscope Shelf.
特徴および利点
Live cell imaging of samples in incubator, remote viewing
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
BMSH356545LCI | 4065269202141 |