物質
( PLA (polylactic acid) resin )
包装
case of 500
メーカー/製品名
Bel-Art F36931-0001
L × W × H
5 3/4 in. × 3/4 in. × 1/16 in.
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Lessen environmental impact with these PLA (polylactic acid) resin sampling sticks. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Whether you are mixing samples, handling capsules, scraping containers or positioning solid samples for examination, 1001 Sticks are ready to assist! Use a separate stick for each material sampled to avoid cross contamination.
特徴および利点
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
Comparable in strength and durability to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
法的情報
Bel-Art is a registered trademark of SP Scienceware, a division of SP
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
BAF369310001-500EA | 4061832259543 |