Key Documents
AXYPDW500C
ディープウェルプレート
for SBS FootPrint, size 96 wells, size 600 μL, pkg of 10x5plates/cs
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121800
NACRES:
NB.22
物質
conical bottom wells
メーカー/製品名
Axygen Inc P-DW-500-C
Corning
包装
pkg of 10x5plates/cs
サイズ
600 μL
96 wells
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
法的情報
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.