主要文書
MAVFLTVP2
Millipore
MAS-100 VF® Portable Air Sampler Validation Protocol
for use with MAS-100 VF® Microbial Air Samplers
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41116500
NACRES:
JA.11
利用可能性
available only in North America
適合性
for use with MAS-100 VF® Microbial Air Samplers
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
MAS-100 VF® Validation Protocol(Letter Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of MAS-100 VF® microbial air sampling systems. The protocol contains an exhaustive list of tests to cover a batch of units to be validated, the test record protocol MAVFLTTR1 including test descriptions and test records must be ordered separately. The protocol follows GMP, EP, USP and JP regulatory guidelines. Only available in North America.
アプリケーション
MAS-100 VF® Validation Protocol is used to validate microbial air sampling system with ease and consistent performance while following regulatory guidelines.
法的情報
MAS-100 VF is a registered trademark of MBV AG