Merck
Z743180
Julabo® 水槽カバー
lift-up gable cover to fit CORIO CD-B33
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41103706
NACRES:
NB.77
包装
pack of 1 ea
メーカー/製品名
Julabo 9970257
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The hinged stainless steel bath lid reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath.
法的情報
Julabo is a registered trademark of Julabo USA, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)