詳細
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini mixers. Horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
法的情報
Rotobot is a trademark of Benchmark Scientific, Inc.
付属品
製品番号
詳細
価格
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
Z742303-1EA | 4061826163047 |