Merck
Z341657
3M 6000/7000シリーズ フィルター
A2 Organic Vapour filter, pkg of 2 ea
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
46182002
NACRES:
NB.35
詳細
Protects against organic vapours with b.p. >65°C up to 10 × OEL* or 5000ppm*, EN141
包装
pkg of 2 ea
メーカー/製品名
3M 6055
利用可能性
not available in USA and Canada
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Gas and vapour filters
Two filters required per respirator.
特徴および利点
このフィルターは3M社の高度なフィルターテクノロジーを採用しているため、呼吸の抵抗を最小限に抑え、快適に使用することができます。保護の必要性に合わせて、ガスフィルターや蒸気フィルター、微粒子フィルターからお選びいただけます。すべてのフィルターは、バヨネット接続により、6000シリーズと7000シリーズの両方に対応します。レスピレーター1つにつき2枚のフィルターが必要になります。
警告
All these filters should only be used for protection against hazards with good warning properties, e.g. smell or taste, and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH). *Where concentrations are given in terms of the OEL or ppm, the filter should only be used for protection against concentrations below the lower value.
法的情報
3M is a trademark of 3M Company
最新バージョンのいずれかを選択してください:
試験成績書(COA)