Merck
HAM58381-01
Luer lock conversion kit, for Hamilton® Microlab® 600 Concorde convertible hand tip probe
pack of 1 kit
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
包装
pack of 1 kit
詳細
The Luer Lock Conversion Kit is an upgrade for the basic Concorde Convertible Tip Hand Probe. This kit converts the Concorde CT to a TLL probe. It does not include a CT probe.
法的情報
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
最新バージョンのいずれかを選択してください:
試験成績書(COA)