Merck
ESYNTH003
Sigma-Aldrich
SynLectro 50 mL Glass Cell with PTFE stopper
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
81141502
NACRES:
NB.61
関連するカテゴリー
詳細
SynLectro 50 mL Glass Cell with PTFE Stopper is the part of the SynLectro Electrolysis Platform for synthetic electrochemistry.
Features and Benefits:
  • Designed to simplify setup, optimize reactivity, and improve safety
  • Side-arm enables connection to reflux condenser or inert gas
  • Chemical resistant material (PTFE)
  • Protects electrical contacts from electrolytes
  • Sleeve (optional) prevents seized joints
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
法的情報
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
最新バージョンのいずれかを選択してください:
試験成績書(COA)