Merck
DWK218065102
DURAN® graduated amber laboratory bottle
new
bottle capacity 750mL, GL 45 thread, without screw cap and pouring ring, borosilicate glass 3.3 bottle
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121818
物質
amber
borosilicate glass 3.3 bottle
ジョイント
joint threaded neck
無菌性
non-sterile
特徴
autoclavable
graduated
without cap
without screw cap and pouring ring
包装
pack of 10 ea
メーカー/製品名
DWK Life Sciences DURAN® 218065102
ボトルの最大容量
750mL
ボトル 直径× H
95 mm × 203 mm
フィッティング
GL 45 thread
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, amber, USP/EP Type I DURAN® borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The amber bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN® Original amber bottles are supplied with or without iconic blue polypropylene cap and drip-free pouring ring for use up to 140 °C. All components are fully autoclavable at 121 or 134 °C.
アプリケーション
DURAN® graduated laboratory bottles are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.
特徴および利点
  • Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
  • The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
  • Very durable and chemically resistant amber colour
  • Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
  • Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
法的情報
DURAN is a registered trademark of DWK Life Sciences
最新バージョンのいずれかを選択してください:
試験成績書(COA)