Merck
CAD7953
Laboratory pipetting needle with 90° blunt ends
gauge 17, L 4 in., 304 stainless steel hub
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122002
NACRES:
NB.03
物質
304 stainless steel hub
包装
pack of 12 ea
メーカー/製品名
Cadence Science 7953
L
4 in.
ゲージ
17
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Pipetting needles are available in a wide choice of sizes for use in laboratory procedures. All needles are made with type 304 stainless steel cannula.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.
最新バージョンのいずれかを選択してください:
試験成績書(COA)