包装
pack of 5 ea
メーカー/製品名
Cadence Science 6523
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
法的情報
Micro-Mate is a registered trademark of Cadence Science
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
CAD6523-5EA | 4061832517902 |